Merck
  • Occurrence of pharmaceutical compounds and pesticides in aquatic systems.
Occurrence of pharmaceutical compounds and pesticides in aquatic systems.
Marine pollution bulletin (2015-05-23)
Maria Gonzalez-Rey, Nathalie Tapie, Karyn Le Menach, Marie-Hélène Dévier, Hélène Budzinski, Maria João Bebianno
ABSTRACT
This paper deals with the detection and quantification of APIs and other priority substances in the Arade River estuary (Portugal) providing the usefulness of Polar Organic Compound Integrative Samplers (POCIS). Thirteen APIs were detected whose variation was site and time dependent. Caffeine was at the highest concentration (804±209 ng/L) followed by theophylline (184±44 ng/L). Other APIs were analgesic, anticonvulsant, non-steroidal anti-inflammatory drugs, anti-lipidemic, anxiolytic and antidepressants. Twenty pesticides comprising atrazine, diuron, isoproturon, terbutryn and simazine included in the Water Framework Directive priority list were also site and time dependent. Carbendazim occurred at the highest concentration (45±18 ng/L at site 1) but atrazine, diuron, isoproturon and simazine levels were below the Environmental Quality Standards. Although the summer impact was unclear, the results highlighted POCIS suitability for profiling these contaminants. This is to our knowledge the first study concerning APIs and pesticides in this area.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis