Merck
  • Vpx-Independent Lentiviral Transduction and shRNA-Mediated Protein Knock-Down in Monocyte-Derived Dendritic Cells.
Vpx-Independent Lentiviral Transduction and shRNA-Mediated Protein Knock-Down in Monocyte-Derived Dendritic Cells.
PloS one (2015-07-25)
Wojciech Witkowski, Jolien Vermeire, Alessia Landi, Evelien Naessens, Hanne Vanderstraeten, Hans Nauwynck, Herman Favoreel, Bruno Verhasselt
ABSTRACT
The function of dendritic cells (DCs) in the immune system is based on their ability to sense and present foreign antigens. Powerful tools to research DC function and to apply in cell-based immunotherapy are either silencing or overexpression of genes achieved by lentiviral transduction. To date, efficient lentiviral transduction of DCs or their monocyte derived counterparts (MDDCs) required high multiplicity of infection (MOI) or the exposure to the HIV-2/SIV protein Vpx to degrade viral restriction factor SAM domain and HD domain-containing protein 1 (SAMHD1). Here we present a Vpx-independent method for efficient (>95%) transduction of MDDCs at lower MOI. The protocol can be used both for ectopic gene expression and knock-down. Introducing shRNA targeting viral entry receptor CD4 and restriction factor SAMHD1 into MDDCs resulted in down-regulation of targeted proteins and, consequently, expected impact on HIV infection. This protocol for MDDCs transduction is robust and free of the potential risk arising from the use of Vpx which creates a virus infection-prone environment, potentially dangerous in clinical setting.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
MISSION® esiRNA, targeting human CD4