Merck
  • Overexpression of CD9 correlates with tumor stage and lymph node metastasis in esophageal squamous cell carcinoma.
Overexpression of CD9 correlates with tumor stage and lymph node metastasis in esophageal squamous cell carcinoma.
International journal of clinical and experimental pathology (2015-06-06)
Jian Huan, Yi Gao, Jing Xu, Wenjiong Sheng, Wei Zhu, Shuyu Zhang, Jianping Cao, Jiang Ji, Liyuan Zhang, Ye Tian
ABSTRACT
Esophageal squamous cell carcinoma (ESCC) is one of the leading causes of cancer deaths worldwide. CD9 has been reported to play a critical role in cell motility, growth and metastasis of multiple cancers. The present study investigated the clinicopathological features of CD9, and its biological characteristics in ESCC. Fifteen normal esophageal tissue specimens, fifty-three ESCC adjacent tissues and one hundred and four ESCC tissues were included in this study. Using immunohistochemistry (IHC), the expression levels of CD9 were evaluated among different samples. And its clinicopathological parameters and its prognostic factors were analyzed. Western blotting was used to measure CD9 expression and colony formation was performed to determine the effect of CD9 on cell growth in ESCC TE-1 cells. Compared with normal esophageal tissues and tumor adjacent tissues, CD9 expression level is significantly higher in ESCC tissues. CD9 expression correlated with tumor stage (P=0.022) and lymph node metastasis (P=0.019) in ESCC patients. Furthermore, the small interfering RNA-mediated silencing of CD9 expression in TE-1 cells resulted in increased proliferation as evidenced by increased colony number and colony size. CD9 expression is upregulated in ESCC tissues and its expression is correlated with tumor stage and lymph node metastasis in ESCC patients. CD9 suppresses the proliferation of TE-1 cells. CD9 may present a potential in tumor progression in ESCC.
Product Description
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
MISSION® esiRNA, targeting human CD9
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cd9
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%