Merck
Novel IL-2-Poly(HPMA)Nanoconjugate Based Immunotherapy.
Journal of biomedical nanotechnology (2015-10-22)
Petra Votavova, Jakub Tomala, Vladimir Subr, Jiri Strohalm, Karel Ulbrich, Blanka Rihova, Marek Kovar
ABSTRACT
Interleukin-2 (IL-2) possesses a strong stimulatory activity for activated T and NK cells and it is an attractive molecule for immunotherapy. Nevertheless, extremely short half-life and severe toxicities associated with high-dose IL-2 treatment are serious and limiting drawbacks. In order to increase IL-2 half-life in vivo, we covalently conjugated synthetic semitelechelic polymeric carrier based on N-(2-hydroxypropyl)methacrylamide (HPMA) to IL-2. Thus, we synthesized IL-2-poly(HPMA) conjugate containing 2-3 polymer chains per IL-2 molecule in average. Such conjugate has lower biologic activity in comparison to IL-2 in vitro. However, it exerts much higher activity than IL-2 in vivo as shown by expansion of memory CD8+ T, NK, NKT, γδT and Treg cells. Moreover, IL-2-poly(HPMA) extremely effectively potentiates CD8+ T cell peptide-based vaccination. IL-2-poly(HPMA) shows also much longer half-time in circulation than IL-2 (-4 h versus -5 min). Collectively, modification of IL-2 with poly(HPMA) chains dramatically improves its potency and pharmacologic features in vivo, which have implications for immunotherapy. To our knowledge, this is the first proof-of-concept report of the use of polymer/protein modification of IL-2 to obtain more pronounced biological activity.
Sigma-Aldrich
Amino-2-propanol, 93%
Sigma-Aldrich
DCC, 99%
Sigma-Aldrich
Methacryloyl chloride, 97%, contains monomethyl ether hydroquinone as stabilizer
Sigma-Aldrich
DCC, 1.0 M in methylene chloride
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Phthaldialdehyde, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Phthaldialdehyde, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
Phthaldialdehyde, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
Octylphosphonic acid, 97%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
2-Thiazoline-2-thiol, 98%