Merck
Urinary cysteinyl progestogens: Occurrence and origin.
The Journal of steroid biochemistry and molecular biology (2015-04-29)
Josep Marcos, Marta Pol, Andreu Fabregat, Rosa Ventura, Nuria Renau, Felicia A Hanzu, Gregori Casals, Santi Marfà, Bernardí Barceló, Antonia Barceló, Juan Robles, Jordi Segura, Oscar J Pozo
ABSTRACT
The presence of two cysteinyl progestogens, 16-cysteinyl-progesterone (16-Cys-Prog) and 16-cysteinyl-pregnenolone (16-Cys-Preg), in human urine is described for the first time. Their occurrence was unequivocally confirmed by comparison with synthesized material by using mass spectrometric detectors. Several experiments were performed in order to clarify their origin. The adrenal origin of both 16-Cys-Prog and 16-Cys-Preg can be inferred from the increase in their concentrations after ACTH stimulatory test, together with their circadian variation similar to the one observed for cortisol. Moreover, the notable increase in excretions of 16-Cys-Prog during the luteal phase of the menstrual cycle points towards an ovarian production for this progestogen. However, the analysis of samples during the course of two pregnancies revealed that, in spite of the large amounts of progesterone produced during gestation, the human placenta lacks the capacity to make 16-Cys-Prog. The adrenal and ovarian origin has been further indicated by the absence of both metabolites in samples collected from a subject with bilateral adrenalectomy and hypogonadotrophyic hypogonadism. Regarding liver action, in vitro studies with hepatocytes and progesterone indicate that, although the liver is able to metabolize progesterone to 6-dehydroprogesterone, it has not the enzymatic machinery for the generation of 16-dehydroprogesterone. Taken together, these results open the possibility for a noninvasive test for the simultaneous evaluation of progesterone biosynthesis in different organs.
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Ammonium iodide, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Supelco
N-Methyl-bis(trifluoroacetamide), derivatization grade (GC derivatization), LiChropur, ≥97.0% (GC)
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Potassium carbonate, powder, 99.99% trace metals basis
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis