Merck
  • Combined platelet and plasma derivatives enhance proliferation of stem/progenitor cells maintaining their differentiation potential.
Combined platelet and plasma derivatives enhance proliferation of stem/progenitor cells maintaining their differentiation potential.
Cytotherapy (2015-11-22)
Anita Muraglia, Maria Rosa Todeschi, Andrea Papait, Alessandro Poggi, Raffaele Spanò, Paolo Strada, Ranieri Cancedda, Maddalena Mastrogiacomo
ABSTRACT
Platelet derivatives have been proposed as alternatives to animal sera given that for cell therapy applications, the use of fetal bovine/calf serum (FBS/FCS) is subjected to severe limitations for safety and ethical concerns. We developed a cell culture medium additive obtained by the combination of two blood-derived standardized components. A platelet lysate (PL) and a platelet-poor plasma (PPP) were produced in a lyophilized form. Each component was characterized for its growth factor content (platelet-derived growth factor-BB/vascular endothelial growth factor). PL and PPP were used as single components or in combination in different ratio at cumulative 5% final concentration in the culture medium. The single components were less effective than the component combination. In primary cell cultures (bone marrow stromal cells, adipose derived adult stem cells, osteoblasts, chondrocytes, umbilical cord-derived mesenchymal stromal cells, lymphocytes), the PL/PPP supplement promoted an increased cell proliferation in respect to the standard FCS culture in a dose-dependent manner, maintaining the cell functionality, clonogenicity, phenotype and differentiative properties throughout the culture. At a different component ratio, the supplement was also used to support proliferation of a cell line (U-937). The PL/PPP supplement is an efficient cell culture medium additive that can replace FCS to promote cell proliferation. It can outdo FCS, especially when adopted in primary cultures from tissue biopsies. Moreover, the dual component nature of the supplement allows the researcher to determine the more appropriate ratio of the two components for the nutritional and functional requirements of the cell type of interest.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Picropodophyllotoxin, ≥96% (HPLC), powder
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis