Merck
  • Comparing mouse and human pluripotent stem cell derived cardiac cells: Both systems have advantages for pharmacological and toxicological screening.
Comparing mouse and human pluripotent stem cell derived cardiac cells: Both systems have advantages for pharmacological and toxicological screening.
Journal of pharmacological and toxicological methods (2015-05-10)
E L Lagerqvist, B A Finnin, D A Elliott, D J Anderson, S M Wu, C W Pouton, J M Haynes
ABSTRACT
Pluripotent stem cells offer an unparalleled opportunity to investigate cardiac physiology, pharmacology, toxicology and pathophysiology. In this paper we describe the use of both mouse (Nkx2-5(eGFP/w)) and human (NKX2-5(eGFP/w)) pluripotent stem cell reporter lines, differentiated toward cardiac lineage, for live single cell high acquisition rate calcium imaging. We also assess the potential of NKX2-5(eGFP/w) cardiac lineage cells for use toxicological screening as well as establish their sensitivity to a shift between low and high oxygen environments. Differentiated mouse Nkx2-5(eGFP/w) cells demonstrated a wide range of spontaneous oscillation rates that could be reduced by ryanodine (10μM), thapsigargin (1μM) and ZD7288 (10μM). In contrast human NKX2-5(eGFP/w) cell activity was only reduced by thapsigargin (1μM). Human cell survival was sensitive to the addition of trastuzumab and doxorubicin, while the switch from a low to a high oxygen environment affected oscillation frequency. We suggest that the human NKX2-5(eGFP/w) cells are less suitable for studies of compounds affecting cardiac pacemaker activity than mouse Nkx2-5(eGFP/w) cells, but are very suitable for cardiac toxicity studies.
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%
Sigma-Aldrich
NNC 55-0396 hydrate, ≥98% (HPLC)
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, natural, 97%, FG