Merck
  • RhoE is required for contact inhibition and negatively regulates tumor initiation and progression.
RhoE is required for contact inhibition and negatively regulates tumor initiation and progression.
Oncotarget (2015-06-04)
Marta Hernández-Sánchez, Enric Poch, Rosa M Guasch, Joaquín Ortega, Inmaculada López-Almela, Ignacio Palmero, Ignacio Pérez-Roger
ABSTRACT
RhoE is a small GTPase involved in the regulation of actin cytoskeleton dynamics, cell cycle and apoptosis. The role of RhoE in cancer is currently controversial, with reports of both oncogenic and tumor-suppressive functions for RhoE. Using RhoE-deficient mice, we show here that the absence of RhoE blunts contact-inhibition of growth by inhibiting p27Kip1 nuclear translocation and cooperates in oncogenic transformation of mouse primary fibroblasts. Heterozygous RhoE+/gt mice are more susceptible to chemically induced skin tumors and RhoE knock-down results in increased metastatic potential of cancer cells. These results indicate that RhoE plays a role in suppressing tumor initiation and progression.
Sigma-Aldrich
MISSION® esiRNA, targeting human RND3
MCF7 Cell Line human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 86012803, human breast (adenocarcinoma), Epithelial-like