Merck
  • Phthalimido-ferrocidiphenol cyclodextrin complexes: Characterization and anticancer activity.
Phthalimido-ferrocidiphenol cyclodextrin complexes: Characterization and anticancer activity.
International journal of pharmaceutics (2015-07-03)
Feten Najlaoui, Pascal Pigeon, Zaineb Abdelkafi, Sebastien Leclerc, Pierrick Durand, Mohamed El Ayeb, Naziha Marrakchi, Ali Rhouma, Gérard Jaouen, Stéphane Gibaud
ABSTRACT
Several ferrocenyl analogues of tamoxifen have already showed strong antiproliferative activity in experimental glioma models. Nevertheless, these compounds are very poorly soluble in water and an adapted formulation is needed. In this work, we have tailored and optimized methylated cyclodextrin soluble complexes of phthalimido-ferrocidiphenol for the first time. The complexes were characterized, and the optimized formulation was tested for in vitro efficacy and cell proliferation assays on U87, human glioblastoma cancer cells. Molecular modeling can provide accurate information about the inclusion process. The inclusion of all the moieties at the same time (i.e., ferrocene, phthalimidylpropyl, 2 phenols) is not possible due to the steric hindrance of the 1:4 system. The 1:3 systems are possible but do not seem very relevant. However, various 1:2 and 1:1 complexes are mostly present in aqueous solutions. Some experiments have confirmed our hypothesis. First, interactions between the phenol, phthalimidylpropyl and ferrocenyl groups have been observed in our NMR experiments. Second, the inclusion of phthalimidylpropyl was detected by UV-vis spectrophotometry with an apparent 1:1 interaction, which was observed through the Benesi-Hildebrand method. The complex is readily soluble in water and keeps its pharmacological activity against U87 tumor cells (IC50=0.028 ± 0.007 μM vs. 0.018 ± 0.003 μM for PhtFerr).
Sigma-Aldrich
γ-Cyclodextrin, Wacker Chemie AG, 98.3-102.0% cyclodextrin basis
Sigma-Aldrich
Titanium tetrachloride, packaged for use in deposition systems
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
γ-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
γ-Cyclodextrin, ≥98%
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Phthalimide, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
γ-Cyclodextrin, Wacker Chemie AG, ≥90.0% cyclodextrin basis (HPLC)
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D