Merck
  • LRG1 expression indicates unfavorable clinical outcome in hepatocellular carcinoma.
LRG1 expression indicates unfavorable clinical outcome in hepatocellular carcinoma.
Oncotarget (2015-10-31)
Chun-Hua Wang, Min Li, Li-Li Liu, Ruo-Yao Zhou, Jia Fu, Chris Zhiyi Zhang, Jing-Ping Yun
ABSTRACT
Leucine-rich-alpha-2-glycoprotein1 (LRG1) is a novel oncogene-associated protein which has been clarified vital to the progression of human cancers, but its role in hepatocellular carcinoma (HCC) remains unclear. Here, we showed that the expression of LRG1 was noticeably increased in HCC tissues, compared to the nontumorous tissues. High LRG1 expression was significantly associated with tumor size (P = 0.004), tumor differentiation (P = 0.010), TNM stage (P < 0.001) and vascular invasion (P = 0.019). Kaplan-Meier analysis showed that LRG1 expression was closely correlated to overall survival and disease-free survival in a training cohort of 474 patients with HCC. The correlation was further validated in an independent cohort of 303 HCC patients. The prognostic implication of LRG1 was confirmed by stratified survival analyses. Multivariate Cox regression model indicated LRG1 as an independent poor prognostic indicator for overall survival (Hazard ratio = 1.582, 95% confident interval: 1.345-1.862, P < 0.001) and disease-free survival (Hazard ratio = 1.280, 95% confident interval: 1.037-1.581, P = 0.022) in HCC. In vitro data showed that LRG1 markedly promoted cell migration but has no effect on cell proliferation. Collectively, our data show that LRG1 is markedly up-regulated and serves as an independent factor of poor outcomes in HCC. Our study therefore provides a promising biomarker for prognostic prediction in clinical management of HCC.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O