Merck
  • Acousto-optic tunable filter near-infrared spectroscopy for in-line monitoring liquid-liquid extraction of Gardenia jasminoides Ellis based on statistical analysis.
Acousto-optic tunable filter near-infrared spectroscopy for in-line monitoring liquid-liquid extraction of Gardenia jasminoides Ellis based on statistical analysis.
Die Pharmazie (2015-11-26)
Sha Wu, Ye Jin, Qi-An Liu, Jian-Xiong Wu, Yu-An Bi, Zhen-Zhong Wang, Wei Xiao
ABSTRACT
This study aimed to monitor liquid-liquid extraction of Gardenia jasminoides Ellis (Zhizi in Chinese) using in-line near-infrared spectroscopy. Shanzhiside (SZS), deacetyl asperulosidic acid methyl ester (DAAME), genipin-1-β-D-gentiobioside (GG), geniposide (GS), total acids (TA) and soluble solid content (SSC) were selected as quality control indicators, and measured by reference methods. Both partial least-squares regression (PLSR) and back propagation artificial neural networks (BP-ANN) were applied to create models to predict the content of above indicators. Paired-samples t-test and nonparametric test were used to compare differences in predictive values between two models of each indicator. Relative standard error of prediction (RSEP) and mean absolute percentage error (MAPE) were used to evaluate the predictive accuracy of the established models. The results showed that there was no significant difference in predicting DAAME, GS and TA between two models. However, PLSR model gave better accuracy in predicting GG and SZS than BP-ANN model. The BP-ANN model of SSC was better than PLSR model. This study shows that NIR spectroscopy can be used for rapid and accurate analysis of quality control indicators in the liquid-liquid extraction of Zhizi. Simultaneously, this study can serve as technical support for the application of NIR spectroscopy in the industrial production process.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O