Merck
  • The Cloud Point of Alkyl Ethoxylates and Its Prediction with the Hydrophilic-Lipophilic Difference (HLD) Framework.
The Cloud Point of Alkyl Ethoxylates and Its Prediction with the Hydrophilic-Lipophilic Difference (HLD) Framework.
Langmuir : the ACS journal of surfaces and colloids (2015-10-16)
Silvia Zarate-Muñoz, Americo Boza Troncoso, Edgar Acosta
ABSTRACT
The hydrophobicity of surfactants has been described through different concepts used to guide the formulation of surfactant-water (SW) and surfactant-oil-water (SOW) systems. An integrated framework of hydrophobicity indicators could provide a complete tool for surfactant characterization, and insights on how their relationship may influence the overall phase behavior of the system. The hydrophilic-lipophilic difference (HLD) and the characteristic curvature (Cc) parameter, included in the HLD, have been shown to correlate with different hydrophobicity indicators including the hydrophilic-lipophilic balance (HLB), packing factor (Pf), phase inversion temperature (PIT), spontaneous curvature (Ho), surfactant partition (K(o-w)), and the critical micelle concentration (CMC). This work aims to investigate whether the HLD can further describe a concomitant hydrophobicity parameter, the cloud point (CP) of alkyl ethoxylates. After applying group contribution models to calculate the Cc of monodisperse (pure) nonionic alkyl ethoxylates, a linear correlation between the calculated Cc and the CP was observed for pure surfactants with 8 ethylene oxide (EO) units or less. Furthermore, using an apparent equivalent alkane carbon number (EACN) to represent the hydrophobicity of the micelle core, the HLD equation was capable of predicting cloud point temperatures of pure alkyl ethoxylates, typically within 5 °C. Polydisperse surfactants did not follow the linear CP-Cc correlation found for pure surfactants. After treating polydisperse samples using a liquid-liquid extraction procedure used to remove the most hydrophobic components in the mixture, the resulting treated surfactants fell in the correlation line of pure alkyl ethoxylates. A closer look at the partition behavior of these treated surfactants showed that their partition, Cc and cloud point are dominated by the most abundant ethoxymers in the treated surfactant. The HLD also predicted the cloud point depression of treated surfactants with increasing sodium chloride concentration. This work shows how the HLD framework could be extended to predict the behavior of SW systems.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
SAFC
Decahydronaphthalene, mixture of cis + trans, 97%
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Decahydronaphthalene, mixture of cis + trans, anhydrous, ≥99%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Decahydronaphthalene, mixture of cis + trans, reagent grade, 98%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Cyclohexane, ACS reagent, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)