Merck
  • Novel all trans-retinoic Acid derivatives: cytotoxicity, inhibition of cell cycle progression and induction of apoptosis in human cancer cell lines.
Novel all trans-retinoic Acid derivatives: cytotoxicity, inhibition of cell cycle progression and induction of apoptosis in human cancer cell lines.
Molecules (Basel, Switzerland) (2015-05-12)
Ebtesam Saad Al-Sheddi, Mai Mohammad Al-Oqail, Quaiser Saquib, Maqsood Ahmed Siddiqui, Javed Musarrat, Abdulaziz Ali Al-Khedhairy, Nida Nayyar Farshori
ABSTRACT
Owing to the pharmacological potential of ATRA (all trans-retinoic acid), a series of retinamides and a 1-(retinoyl)-1,3-dicyclohexylurea compound were prepared by reacting ATRA with long chain alkyl or alkenyl fatty amines by using a 4-demethylaminopyridine (DMAP)-catalyzed N,N¢-dicyclohexylcarbodiimide (DCC) coupling. The successful synthesis of the target compounds was demonstrated using a range of spectroscopic techniques. The cytotoxicity of the compounds was measured along with their ability to induce cell cycle arrest and apoptosis in human cancer cell lines MCF-7 (breast cancer) and HepG2 (liver cancer) and normal human cell line HEK293 (embryonic kidney). The results of cytotoxicity and flow cytometry data showed that the compounds had a moderate to strong effect against MCF-7 and HepG2 cells and were less toxic to HEK293 cells. N-oleyl-retinamide was found to be the most potent anticancer agent and was more effective against MCF-7 cells than HepG2 cells.
Sigma-Aldrich
Octadecylamine, ≥99.0% (GC)
Sigma-Aldrich
Octadecylamine, ≥99% (GC)
Sigma-Aldrich
Octadecylamine, technical grade, 90%
SAFC
Oleylamine, ≥98% (primary amine)
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
DCC, 99%
Sigma-Aldrich
DCC, 1.0 M in methylene chloride
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%