Merck
  • Highly selective enrichment of baicalin in rat plasma by boronic acid-functionalized core-shell magnetic microspheres: Validation and application to a pharmacokinetic study.
Highly selective enrichment of baicalin in rat plasma by boronic acid-functionalized core-shell magnetic microspheres: Validation and application to a pharmacokinetic study.
Talanta (2015-11-26)
Taomin Huang, Ya Xiong, Nianzu Chen, Donglei Wang, Yonghua Lai, Chunhui Deng
ABSTRACT
To the best of our knowledge, this study is the first to successfully apply a novel, highly selective enrichment technique based on boronic acid-functionalized core-shell magnetic microspheres (BA-Fe3O4@SiO2-Au@mSiO2) with a large surface area and uniform pore size, to determine the baicalin concentration in rat plasma by HPLC. By taking advantage of the special interaction between boronic acid and baicalin under alkaline conditions, as well as the microspheres' size exclusion ability, baicalin was selectively extracted from protein-rich biosamples, such as plasma, without any other pretreatment procedure except for a 10-min vortexing step. BA-Fe3O4@SiO2-Au@mSiO2 microsphere-adsorbed baicalin was straightforwardly and rapidly isolated from the matrix using a magnet. Baicalin was subsequently eluted from the microspheres under acidic conditions for 2min for further HPLC analysis. The extraction conditions, such as the amount of microspheres added, adsorption time, adsorption pH, and elution time and pH, were also determined. Furthermore, method validation, including the linear range, detection limit, precision, accuracy, and recovery, were determined. This newly developed method based on BA-Fe3O4@SiO2-Au@mSiO2 microspheres is a simple, accurate, selective, and green analytical preparatory technique for analyzing baicalin in rat plasma. This study will be further novel research on the analysis of complex plasma samples and the pharmacokinetics of drugs similar to baicalin.
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt, ≥96.0% (HPCE), solid
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, BioReagent, ≥96.0% (HPCE)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Butylamine, 99.5%
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O