Merck
  • Antibiofilm Activity of Chilean Propolis on Streptococcus mutans Is Influenced by the Year of Collection.
Antibiofilm Activity of Chilean Propolis on Streptococcus mutans Is Influenced by the Year of Collection.
BioMed research international (2015-08-08)
Jorge Jesús Veloz, Nicolás Saavedra, Alexis Lillo, Marysol Alvear, Leticia Barrientos, Luis A Salazar
ABSTRACT
The chemical composition of propolis varies according to factors that could have an influence on its biological properties. Polyphenols from propolis have demonstrated an inhibitory effect on Streptococcus mutans growth. However, it is not known if different years of propolis collection may affect its activity. We aimed to elucidate if the year of collection of propolis influences its activity on Streptococcus mutans. Polyphenol-rich extracts were prepared from propolis collected in three different years, characterized by LC-MS and quantified the content of total polyphenols and flavonoids groups. Finally, was evaluated the antibacterial effect on Streptococcus mutans and the biofilm formation. Qualitative differences were observed in total polyphenols, flavones, and flavonols and the chemical composition between the extracts, affecting the strength of inhibition of biofilm formation but not the antimicrobial assays. In conclusion, chemical composition of propolis depends on the year of collection and influences the strength of the inhibition of biofilm formation.
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Sigma-Aldrich
Caffeic acid phenethyl ester, ≥97% (HPLC), powder
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder