Merck
  • An ultrahigh-performance liquid chromatography method with electrospray ionization tandem mass spectrometry for simultaneous quantification of five phytohormones in medicinal plant Glycyrrhiza uralensis under abscisic acid stress.
An ultrahigh-performance liquid chromatography method with electrospray ionization tandem mass spectrometry for simultaneous quantification of five phytohormones in medicinal plant Glycyrrhiza uralensis under abscisic acid stress.
Journal of natural medicines (2015-02-25)
Yu Xiang, Xiaona Song, Jing Qiao, Yimei Zang, Yanpeng Li, Yong Liu, Chunsheng Liu
ABSTRACT
An efficient simplified method was developed to determine multiple classes of phytohormones simultaneously in the medicinal plant Glycyrrhiza uralensis. Ultrahigh-performance liquid chromatography electrospray ionization tandem mass spectrometry (UPLC/ESI-MS/MS) with multiple reaction monitoring (MRM) in negative mode was used for quantification. The five studied phytohormones are gibberellic acid (GA3), abscisic acid (ABA), jasmonic acid (JA), indole-3-acetic acid, and salicylic acid (SA). Only 100 mg of fresh leaves was needed, with one purification step based on C18 solid-phase extraction. Cinnamic acid was chosen as the internal standard instead of isotope-labeled internal standards. Under the optimized conditions, the five phytohormones with internal standard were separated within 4 min, with good linearities and high sensitivity. The validated method was applied to monitor the spatial and temporal changes of the five phytohormones in G. uralensis under ABA stress. The levels of GA3, ABA, JA, and SA in leaves of G. uralensis were increased at different times and with different tendencies in the reported stress mode. These changes in phytohormone levels are discussed in the context of a possible feedback regulation mechanism. Understanding this mechanism will provide a good chance of revealing the mutual interplay between different biosynthetic routes, which could further help elucidate the mechanisms of effective composition accumulation in medicinal plants.
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Gibberellic acid, 90% gibberellin A3 basis (HPLC)
Sigma-Aldrich
Gibberellic acid, BioReagent, suitable for plant cell culture, ≥90% gibberellin A3 basis (of total gibberellins.)
Sigma-Aldrich
Indole-3-acetic acid sodium salt, suitable for plant cell culture, BioReagent, ≥98%
Sigma-Aldrich
Gibberellic acid potassium salt, suitable for plant cell culture, BioReagent, ~95%, ≥50% total GA3 basis
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Sigma-Aldrich
(+)-Abscisic acid, ≥98% (HPLC)
Sigma-Aldrich
3-Indoleacetic acid, 98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Gibberellin, 80% gibberellin A3 basis (TLC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)