Merck
  • Circulating inflammatory miRNA signature in response to different doses of aerobic exercise.
Circulating inflammatory miRNA signature in response to different doses of aerobic exercise.
Journal of applied physiology (Bethesda, Md. : 1985) (2015-05-23)
David de Gonzalo-Calvo, Alberto Dávalos, Ana Montero, Ángela García-González, Iryna Tyshkovska, Antonio González-Medina, Sara M A Soares, Pablo Martínez-Camblor, Patricia Casas-Agustench, Manuel Rabadán, Ángel E Díaz-Martínez, Natalia Úbeda, Eduardo Iglesias-Gutiérrez
ABSTRACT
While moderate acute exercise has been associated with strong anti-inflammatory mechanisms, strenuous exercise has been linked to deleterious inflammatory perturbations. It is therefore fundamental to elucidate the mechanisms that regulate the exercise-induced inflammatory cascade. Information on novel regulators such as circulating inflammatory microRNAs (c-inflammamiRs) is incomplete. In this study, we evaluated the response of a panel of c-inflammamiRs to different doses of acute aerobic exercise. We first studied the exercise-induced inflammatory cascade in serum samples of nine active middle-aged males immediately before and after (0 h, 24 h, 72 h) 10-km, half-marathon, and marathon races. Next, we analyzed the circulating profile of 106 specific c-inflammamiRs immediately before) and after (0 h, 24 h) 10-km (low inflammatory response) and marathon (high inflammatory response) races. Analysis of classical inflammatory parameters revealed a dose-dependent effect of aerobic exercise on systemic inflammation, with higher levels detected after marathon. We observed an increase in miR-150-5p immediately after the 10-km race. Levels of 12 c-inflammamiRs were increased immediately after the marathon (let-7d-3p, let-7f-2-3p, miR-125b-5p, miR-132-3p, miR-143-3p, miR-148a-3p, miR-223-3p, miR-223-5p, miR-29a-3p, miR-34a-5p, miR-424-3p, and miR-424-5p). c-inflammamiRs returned to basal levels after 24 h. Correlation and in silico analyses supported a close association between the observed c-inflammamiR pattern and regulation of the inflammatory process. In conclusion, we found that different doses of acute aerobic exercise induced a distinct and specific c-inflammamiR response, which may be associated with control of the exercise-induced inflammatory cascade. Our findings point to c-inflammamiRs as potential biomarkers of exercise-induced inflammation, and hence, exercise dose.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Creatine, anhydrous
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder