Merck
  • An Automated Method for the Determination of Trimebutine and N-Mono-Desmethyl Trimebutine by On-Line Turbulent Flow Coupled with Liquid Chromatography-Tandem Mass Spectrometry in Human Plasma: Application to a Fatal Poisoning Case with Toxicokinetic Study.
An Automated Method for the Determination of Trimebutine and N-Mono-Desmethyl Trimebutine by On-Line Turbulent Flow Coupled with Liquid Chromatography-Tandem Mass Spectrometry in Human Plasma: Application to a Fatal Poisoning Case with Toxicokinetic Study.
Journal of analytical toxicology (2015-09-06)
Islam Amine Larabi, Charlotte Duverneuil-Mayer, Emuri Abe, Frédéric Baud, Jean-Claude Alvarez
ABSTRACT
A liquid chromatography-MS-MS turbulent flow on-line extraction method was developed for the determination of trimebutine (TMB) and its main active metabolite N-mono-desmethyltrimebutine (nortrimebutine or nor-TMB) in human plasma. After protein precipitation and internal standard (IS, haloperidol-d4) addition, 50 µL of the supernatant were transferred onto a Cyclone-Turbo-Flow extraction column followed by an Hypersil PFP Gold analytical column. Detection was carried out on a triple quadrupole tandem mass spectrometer using positive electrospray ionization. The transitions used were m/z 388.0→343.0, 374.0→195.0 and 380.1→169.0 for TMB, nor-TMB and IS, respectively. The method was validated over the concentration range of 10-1,000 ng/mL for both compounds. The accuracy evaluated at three concentrations was within 90.0-98.5% and the intra- and interday coefficient of variation's for the two molecules were <8.7%. The method was applied to a toxicokinetic study of a self-poisoning case with TMB in a 19-old girl. The concentration of TMB decreased from 747 to 77 ng/mL, while nor-TMB decreased from 9,745 to 205 ng/mL after 5 days and the fatal issue. This case confirms the literature underlining the potential toxicity of TMB, which has long time been considered as a harmless molecule.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%