Merck
  • A high-efficiency induction of dopaminergic cells from human umbilical mesenchymal stem cells for the treatment of hemiparkinsonian rats.
A high-efficiency induction of dopaminergic cells from human umbilical mesenchymal stem cells for the treatment of hemiparkinsonian rats.
Cell transplantation (2014-10-08)
Tsui-Ling Ko, Yu-Yi Fu, Yang-Hsin Shih, Yi-Hui Lin, Miau-Hwa Ko, Tz-Win Fu, Tzu-Yung Lin, Hsiao-Sheng Hsiao, Pei-Ming Chu, Yu-Show Fu
ABSTRACT
The success rate in previous attempts at transforming human umbilical mesenchymal stem cells (HUMSCs) isolated from Wharton's jelly of the umbilical cord into dopaminergic cells was a mere 12.7%. The present study was therefore initiated to establish a more effective procedure for better yield of dopaminergic cells in such transformation for more effective HUMSC-based therapy for parkinsonism. To examine, in vitro, the effects of enhanced Nurr1 expression in HUMSCs on their differentiation, cells were processed through the three-stage differentiation protocol. The capacity of such cells to synthesize and release dopamine was measured by HPLC. The therapeutic effects of Nurr1-overexppressed HUMSCs were examined in 6-hydroxydopamine-lesioned rats by quantification of rotations in response to amphetamine. Enhanced Nurr1 expression in HUMSCs promoted the transformation into dopaminergic cells in vitro through stepwise culturing in sonic hedgehog, fibroblast growth factor-8, and neuron-conditioned medium. The success rate was about 71%, as determined by immunostaining for tyrosine hydroxylase and around 94 nM dopamine synthesis (intracellular and released into the culture medium), as measured by HPLC. Additionally, transplantation of such cells into the striatum of hemiparkinsonian rats resulted in improvement of their behavioral deficits, as indicated by amphetamine-evoked rotation scores. Viability of the transplanted cells lasted for at least 3 months as verified by positive staining for tyrosine hydroxylase. Nurr1, FGF8, Shh, and NCM can synergistically enhance the differentiation of HUMSCs into dopaminergic cells and may pave the way for HUMSC-based treatments for Parkinson's disease.
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, Rhodamine conjugate, Chemicon®, from goat
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, ascites fluid, clone LNC1, Chemicon®
Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)
Sigma-Aldrich
Serotonin creatinine sulfate monohydrate, powder
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, biotin-SP conjugate, Chemicon®, from goat
Sigma-Aldrich
Perchloric acid, ACS reagent, 70%
Sigma-Aldrich
Perchloric acid, ACS reagent, 60%