Merck
  • Application of chemometrics in determination of the acid dissociation constants (pKa) of several benzodiazepine derivatives as poorly soluble drugs in the presence of ionic surfactants.
Application of chemometrics in determination of the acid dissociation constants (pKa) of several benzodiazepine derivatives as poorly soluble drugs in the presence of ionic surfactants.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2015-01-01)
Tavakol Heidary Shayesteh, Moojan Radmehr, Farzad Khajavi, Reza Mahjub
ABSTRACT
In this study, the acid dissociation constants (pKa) of some benzodiazepine derivatives including chlordiazepoxide, clonazepam, lorazepam, and oxazepam in aqueous micellar solution were determined spectrophotometrically at an ionic strength of 0.1M at 25°C. The effect of cetyl trimethylammonium bromide (CTAB) as a cationic and sodium n-dodecyl sulfate(SDS) as an anionic surfactant on the absorption spectra of benzodiazepine drugs at different pH values were studied. The acidity constants of all related species are estimated by considering the surfactant concept and the application of chemometric methods using the whole spectral fitting of the collected data to an established factor analysis model. DATAN® software (Ver. 5.0, Multid Analyses AB, and Goteborg, Sweden) was applied to determine the acidity constants. In this study, a simple and fast method to determine the ionization constant (pKa) of poorly soluble drugs was developed using surfactants. The acidity constant (i.e. pKa) for chlordiazepoxide, clonazepam, lorazepam, and oxazepam were reported as 4.62, pKa1 value of 1.52 and pKa2 value of 10.51, pKa1 value of 1.53 and pKa2 value of 10.92 and pKa1 value 1.63 and pKa2 value of 11.21 respectively. The results showed that the peak values in the spectrophotometric absorption spectra of drugs are influenced by the presence of anionic and cationic surfactants. According to the results, by changing the SDS concentration from 0 to 0.05M, the pKa of chlordiazepoxide was increased to 5.9, the pKa1 of lorazepam was decreased to 0.1 while the pKa2 was increased to 11.5. Increase in SDS concentration has not shown significant alteration in pKa of clonazepam and oxazepam. Results indicate that by Changing the CTAB concentration from 0 to 0.05M, the pKa of chlordiazepoxide was reduced to 4.4, the pKa1 of clonazepam was decreased to 0.1 and the pKa2 was decreased to 9.1, the pKa1 of lorazepam was decreased to 0.4 and the pKa2 was decreased to 9.4, the pKa1 of oxazepam was decreased to 0.3 and the pKa2 was decreased to 9.7. Based on the results obtained from the study, charge of anionic and cationic surfactants leads to an electrostatic interaction between surfactant and the protonated form of the drug molecule. The electrostatic interactions can be attractive or repulsive forces and influence of separation of protons and consequently increase or decrease the acidity constants.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Potassium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Potassium nitrate, 99.99% trace metals basis
Sigma-Aldrich
Potassium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)