Merck
  • Registered report: Interactions between cancer stem cells and their niche govern metastatic colonization.
Registered report: Interactions between cancer stem cells and their niche govern metastatic colonization.
eLife (2015-06-19)
Francesca Incardona, M Mehdi Doroudchi, Nawfal Ismail, Alberto Carreno, Erin Griner, Minyoung Anna Lim
ABSTRACT
The Reproducibility Project: Cancer Biology seeks to address growing concerns about reproducibility in scientific research by replicating selected results from a substantial number of high-profile papers in the field of cancer biology published between 2010 and 2012. This Registered report describes the proposed replication plan of key experiments from 'Interactions between cancer stem cells and their niche govern metastatic colonization' by Malanchi and colleagues, published in Nature in 2012 (Malanchi et al., 2012). The key experiments that will be replicated are those reported in Figures 2H, 3A, 3B, and S13. In these experiments, Malanchi and colleagues analyze messenger RNA levels of periostin (POSTN) in pulmonary fibroblasts, endothelial cells, and immune cells isolated from mice with micrometastases to determine which cell type is producing POSTN in the metastatic niche (Figure 2H; Malanchi et al., 2012). Additionally, they examine MMTV-PyMT control or POSTN null mice to test the effect of POSTN on primary tumor growth and metastasis (Figures 3A, 3B, and S13; Malanchi et al., 2012). The Reproducibility Project: Cancer Biology is a collaboration between the Center for Open Science and Science Exchange, and the results of the replications will be published in eLife.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Roche
Liberase TH Research Grade, lyophilized, suitable for tissue processing, optimum pH 7.4