Merck
Enhanced Shewanella biofilm promotes bioelectricity generation.
Biotechnology and bioengineering (2015-04-23)
Ting Liu, Yang-Yang Yu, Xiao-Peng Deng, Chun Kiat Ng, Bin Cao, Jing-Yuan Wang, Scott A Rice, Staffan Kjelleberg, Hao Song
ABSTRACT
Electroactive biofilms play essential roles in determining the power output of microbial fuel cells (MFCs). To engineer the electroactive biofilm formation of Shewanella oneidensis MR-1, a model exoelectrogen, we herein heterologously overexpressed a c-di-GMP biosynthesis gene ydeH in S. oneidensis MR-1, constructing a mutant strain in which the expression of ydeH is under the control of IPTG-inducible promoter, and a strain in which ydeH is under the control of a constitutive promoter. Such engineered Shewanella strains had significantly enhanced biofilm formation and bioelectricity generation. The MFCs inoculated with these engineered strains accomplished a maximum power density of 167.6 ± 3.6 mW/m(2) , which was ∼ 2.8 times of that achieved by the wild-type MR-1 (61.0 ± 1.9 mW/m(2) ). In addition, the engineered strains in the bioelectrochemical system at poised potential of 0.2 V vs. saturated calomel electrode (SCE) generated a stable current density of 1100 mA/m(2) , ∼ 3.4 times of that by wild-type MR-1 (320 mA/m(2) ).
Sigma-Aldrich
BES, ≥99.0% (titration)
Sigma-Aldrich
Diethyl azodicarboxylate solution, purum, ~40% in toluene (H-NMR)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
SAFC
Isopropyl β-D-1-thiogalactopyranoside