Merck
  • Penicillin V acylase from Pectobacterium atrosepticum exhibits high specific activity and unique kinetics.
Penicillin V acylase from Pectobacterium atrosepticum exhibits high specific activity and unique kinetics.
International journal of biological macromolecules (2015-05-02)
V S Avinash, Sureshkumar Ramasamy, C G Suresh, Archana Pundle
ABSTRACT
Penicillin V acylases (PVAs, E.C.3.5.11) belong to the Ntn hydrolase super family of enzymes that catalyze the deacylation of the side chain from phenoxymethyl penicillin (penicillin V). Penicillin acylases find use in the pharmaceutical industry for the production of semi-synthetic antibiotics. PVAs employ the N-terminal cysteine residue as catalytic nucleophile and are structurally and evolutionarily related to bile salt hydrolases (BSHs). Here, we report the cloning and characterization of a PVA enzyme from the Gram-negative plant pathogen, Pectobacterium atrosepticum (PaPVA). The enzyme was cloned and expressed in Escherichia coli attaining a very high yield (250 mg/l) and a comparatively high specific activity (430 IU/mg). The enzyme showed marginally better pH and thermo-stability over PVAs characterized from Gram-positive bacteria. The enzyme also showed enhanced activity in presence of organic solvents and detergents. The enzyme kinetics turned out to be significantly different from that of previously reported PVAs, displaying positive cooperativity and substrate inhibition. The presence of bile salts had a modulating effect on PaPVA activity. Sequence analysis and characterization reveal the distinctive nature of these enzymes and underscore the need to study PVAs from Gram-negative bacteria.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Phenoxyacetyl chloride, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Phenoxyacetic acid, 98%
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Guanidine hydrochloride, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
SAFC
Guanidine hydrochloride
Sigma-Aldrich
Guanidine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99%