Merck
  • Antibacterial activity of silver nanoparticles target sara through srna-teg49, a key mediator of hfq, in staphylococcus aureus.
Antibacterial activity of silver nanoparticles target sara through srna-teg49, a key mediator of hfq, in staphylococcus aureus.
International journal of clinical and experimental medicine (2015-07-02)
Hu Tian, Qiande Liao, Meizhou Liu, Jianhong Hou, Yangde Zhang, Ju Liu
ABSTRACT
Attributed to its antimicrobial effect, Silver nanoparticles (AgNPs) is widely used in various fields, such as biomedicine, textiles, health care products and food, etc. However, the antibacterial mechanism of AgNPs in staphylococcus aureus (S. aureus) by regulating sRNA expression remains largely unknown. This study was performed to investigate the involvement of the antibacterial mechanism of AgNPs through sRNA-TEG49, a key mediator of Hfq, in S. aureus. Through the antimicrobial tests of AgNPs, its antibacterial laps and minimum inhibitory concentration was measured. A hierarchical cluster analysis of the differentially expressed sRNA in S. aureus was performed to investigate the relationship between AgNPs and sRNA. Expression of genes was analyzed by real-time PCR. In the present study we found that at the concentrations higher than 1 mg/L, AgNPs could completely restrain bacteria growth, and the antibacterial activity of AgNPs apparently declined at the concentrations lower than 1 mg/L. S. aureus exposure to AgNPs, the expression of sRNA-TEG49, Hfq and sarA was significantly up-regulated in wild-type S. aureus. Moreover, Hfq loss-of-function inhibited the expression of sRNA-TEG49 in mutant-type S. aureus. Furthermore, sRNA-TEG49 loss-of-function associated with down-regulation the expression of sarA in mutant-type S. aureus. It was reasonable that Hfq regulated a distinct underlying molecular and antibacterial mechanism of AgNPs by forming a positive feedback loop with sRNA-TEG49. These observations suggested that Hfq plays an important role in the antibacterial mechanism of AgNPs by regulating sRNA-TEG49 expression, via its target sarA.
Sigma-Aldrich
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
Sigma-Aldrich
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
Sigma-Aldrich
Silver nitrate, BioXtra, >99% (titration)
Sigma-Aldrich
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
Sigma-Aldrich
Silver nitrate, 99.9999% trace metals basis
Sigma-Aldrich
Silver nitrate, ≥99.999% trace metals basis