Merck
  • Automatic in-syringe dispersive liquid-liquid microextraction of ⁹⁹Tc from biological samples and hospital residues prior to liquid scintillation counting.
Automatic in-syringe dispersive liquid-liquid microextraction of ⁹⁹Tc from biological samples and hospital residues prior to liquid scintillation counting.
Analytical and bioanalytical chemistry (2015-05-27)
Marina Villar, Jessica Avivar, Laura Ferrer, Antoni Borràs, Fernando Vega, Víctor Cerdà
ABSTRACT
A new approach exploiting in-syringe dispersive liquid-liquid microextraction (DLLME) for (99)Tc extraction and preconcentration from biological samples, i.e., urine and saliva, and liquid residues from treated patients is presented. (99)Tc is a beta emitter with a long half-life (2.111 × 10(5) years) and mobility in the different environmental compartments. One of the sources of this radionuclide is through the use of its father (99m)Tc in medical diagnosis. For the first time a critical comparison between extractants and disperser solvents for (99)Tc DLLME is presented, e.g., tributyl phosphate (TBP), trioctylmethylammonium chloride (Aliquat®336), triisooctylamine (TiOA), as extractants in apolar solvents such as xylene and dodecane, and disperser solvents such as acetone, acetonitrile, ethanol, methanol, 1-propanol, and 2-propanol. The system was optimized by experimental design, and 22.5% of Aliquat®336 in acetone was selected as extractant and disperser, respectively. Off-line detection was performed using a liquid scintillation counter. The present method has a (99)Tc minimum detectable activity (MDA) of 0.075 Bq with a high extraction/preconcentration frequency (8 h(-1)). Urine, saliva, and hospital residues were satisfactorily analyzed with recoveries of 82-119%. Thus, the proposed system is an automatic powerful tool to monitor the entry of (99)Tc into the environment. Graphical Abstract (99m)Tc is widely used in Nuclear Medicine for diagnosis. Its daugther (99)Tc is automatically monitored in biological samples from treated patients by in-syringe dispersive liquid-liquid microextraction.
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
1,4-Dithioerythritol, BioXtra, ≥99.0%
Sigma-Aldrich
1,4-Dithioerythritol, ≥99.0%
Sigma-Aldrich
1,4-Dithioerythritol, BioReagent, Molecular Biology, ≥99.0%
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
1,4-Dithioerythritol, BioUltra, ≥99.0% (RT)
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Tributylphosphine solution, 200 mM (in N-methyl-2-pyrrolidinone), liquid
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Dodecane, anhydrous, ≥99%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis