Merck
  • Fabrication of Nanoscale Circuits on Inkjet-Printing Patterned Substrates.
Fabrication of Nanoscale Circuits on Inkjet-Printing Patterned Substrates.
Advanced materials (Deerfield Beach, Fla.) (2015-05-27)
Shuoran Chen, Meng Su, Cong Zhang, Meng Gao, Bin Bao, Qiang Yang, Bin Su, Yanlin Song
ABSTRACT
Nanoscale circuits are fabricated by assembling different conducting materials (e.g., metal nanoparticles, metal nano-wires, graphene, carbon nanotubes, and conducting polymers) on inkjet-printing patterned substrates. This non-litho-graphy strategy opens a new avenue for integrating conducting building blocks into nanoscale devices in a cost-efficient manner.
Sigma-Aldrich
8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, BioReagent, ≥96.0% (HPCE)
Sigma-Aldrich
8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt, ≥96.0% (HPCE), solid
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%