Merck
Development and Optimization of Glaser-Hay Bioconjugations.
Angewandte Chemie (International ed. in English) (2015-06-20)
Jessica S Lampkowski, Jordan K Villa, Travis S Young, Douglas D Young
ABSTRACT
The prevalence of bioconjugates in the biomedical sciences necessitates the development of novel mechanisms to facilitate their preparation. Towards this end, the translation of the Glaser-Hay coupling to an aqueous environment is examined, and its potential as a bioorthogonal conjugation reaction is demonstrated. This optimized, novel, and aqueous Glaser-Hay reaction is applied towards the development of bioconjugates utilizing protein expressed with an alkynyl unnatural amino acid. Unnatural amino acid technology provides a degree of bioorthognality and specificity not feasible with other methods. Moreover, the scope of the reaction is demonstrated through protein-small molecule couplings, small-molecule-solid-support couplings, and protein-solid-support immobilizations.
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ReagentPlus®, 99%
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, ≥99.5%, purified by redistillation
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, Molecular Biology, ≥99% (GC)
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%