Merck
Characterization of a naturally occurring truncated Dicer.
Molecular biology reports (2015-04-26)
Nicola Mosca, Julia Starega-Roslan, Filomena Castiello, Aniello Russo, Wlodzimierz J Krzyzosiak, Nicoletta Potenza
ABSTRACT
Dicer is central to small RNA silencing pathways, thus playing an important role in physiological and pathological states. Recently, a number of mutations in dicer gene have been identified in diverse types of cancer, implicating Dicer in oncogenic cooperation. Here we report on the properties of a rare splice variant of the human dicer gene, occurring in neuroblastoma cells, and not detectable in normal tissues. Due to the skipping of one exon, the alternatively spliced transcript encodes a putative truncated protein, t-Dicer, lacking the dsRNA-binding domain and bearing altered one of the two RNase III catalytic centers. The ability of the exon-depleted t-dicer transcript to be translated in vitro was first investigated by the expression of flagged t-Dicer in human cells. We found that t-dicer transcript could be translated in vitro, albeit not as efficiently as full-length dicer transcript. Then, the possible enzymatic activity of t-Dicer was analyzed by an in vitro dicing assay able to distinguish the enzymatic activity of the individual RNase III domains. We showed that t-Dicer preserved partial dicing activity. Overall, the results indicate that t-dicer transcript could produce a protein still able to bind the substrate and to cleave only one of the two pre-miRNA strands. Given the increasing number of mutations reported for dicer gene in tumours, our experimental approach could be useful to characterize the activity of these mutants, which may dictate changes in selected classes of small RNAs and/or lead to their aberrant maturation.
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Tricine, suitable for cell culture, ≥99% (titration)
Sigma-Aldrich
Tricine, ≥99% (titration)
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Tricine, BioXtra, pH 4.0-6.0 (1 M in H2O), ≥99% (titration)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
SAFC
Tricine
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Glycerin, meets USP testing specifications