- Nanoplatforms attached Schiff bases by condensation method; Investigation of glucose oxidase enzyme as biocatalysts.
Nanoplatforms attached Schiff bases by condensation method; Investigation of glucose oxidase enzyme as biocatalysts.
Artificial cells, nanomedicine, and biotechnology (2013-11-22)
Seda Kayhan, Nurşen Sarı, Dilek Nartop
PMID24256136
ABSTRACT
We report a easy approach for the immobilization of glucose oxidase enzyme (GOx) on nanoparticles. Nanoparticles-modified polystyrene-ANH2 (PSA) with some salicylaldehyde derivatives were synthesized by means of condensation and investigated the enzymatic properties of GOx immobilized on there. Modified polystyrenes were characterized using IR spectra, gel permeation chromatography, and scanning electron microscopy. All the characteristics of the immobilized glucose oxidase (PSA-SalH)-GOx, (PSA-SalCH3)-GOx, and (PSA)-GOx enzyme showed except one of them. Immobilized GOx on to (PSA) showed two optimum pH, other immobilized polymers were one optimum.
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology