Merck
Toxicity of hydroxyurea in rats and dogs.
Toxicologic pathology (2014-11-14)
Daniel Morton, Lori Reed, Wenhu Huang, John M Marcek, Robert Austin-LaFrance, Carrie A Northcott, Scott H Schelling, Bradley E Enerson, Lindsay Tomlinson
ABSTRACT
The toxicity of hydroxyurea, a treatment for specific neoplasms, sickle-cell disease, polycythemia, and thrombocytosis that kills cells in mitosis, was assessed in repeat-dose, oral gavage studies in rats and dogs and a cardiovascular study in telemetered dogs. Hydroxyurea produced hematopoietic, lymphoid, cardiovascular, and gastrointestinal toxicity with steep dose response curves. In rats dosed for 10 days, 50 mg/kg/day was tolerated; 500 mg/kg/day produced decreased body weight gain; decreased circulating leukocytes, erythrocytes, and platelets; decreased cellularity of thymus, lymph nodes, and bone marrow; and epithelial degeneration and/or dysplasia of the stomach and small intestine; 1,500 mg/kg/day resulted in deaths on day 5. In dogs, a single dose at ≥ 250 mg/kg caused prostration leading to unscheduled euthanasia. Dogs administered 50 mg/kg/day for 1 month had decreased circulating leukocytes, erythrocytes, and platelets; increased bone marrow cellularity with decreased maturing granulocytes; increased creatinine kinase activity; and increased iron pigment in bone marrow and hepatic sinusoidal cells. In telemetered dogs, doses ≥ 15 mg/kg decreased systolic blood pressure (BP); 50 mg/kg increased diastolic BP, heart rate, and change in blood pressure over time (+dP/dt), and decreased QT and PR intervals and maximum left ventricular systolic and end diastolic pressures with measures returning to control levels within 24 hr.
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Hydroxyurea, 98%, powder