Merck
  • Impact of Precursors Creatine, Creatinine, and Glucose on the Formation of Heterocyclic Aromatic Amines in Grilled Patties of Various Animal Species.
Impact of Precursors Creatine, Creatinine, and Glucose on the Formation of Heterocyclic Aromatic Amines in Grilled Patties of Various Animal Species.
Journal of food science (2015-10-08)
Monika Gibis, Jochen Weiss
ABSTRACT
The impact of precursors such as creatine, creatinine, and glucose on the formation of mutagenic/carcinogenic heterocyclic amines (HAs) were studied in patties of 9 different animal species equally heat treated with a double-plate contact grill. All grilled patties of the various species (veal, beef, pork, lamb, horse, venison, turkey, chicken, ostrich) contained several HAs such as MeIQx (2-amino-3,8-dimethylimidazo[4,5-f] quinoxaline; 0.5-1.4 ng/g), 4,8-DiMeIQx (2-amino-3,4,8-trimethylimidazo[4,5-f] quinoxaline, 0 to 1.3 ng/g), PhIP (2-amino-1-methyl-6-phenylimidazo[4,5-b] pyridine, 1.2 to 10.5 ng/g), harman (1-methyl-9H-pyrido[3,4-b] indole; 0.5 to 3.2 ng/g), and/or norharman (9H-pyrido[3,4-b]indole 0.5 to 1.9 ng/g). Residual glycogen (glucose) content varied greatly from 0.07 to 1.46 wt% on a dry matter (DM) basis. Total creatin(in)e content in raw meat (1.36 to 2.0 wt% DM) hardly differed between species, except in turkey and ostrich (1.1 wt% DM). Chicken contained, compared to all other species, very low concentrations of glucose (0.07 wt% DM) and the highest levels of nonprotein nitrogen compounds. The free amino acids lysine (r = 0.77, P < 0.001), tyrosine, phenylalanine, proline, isoleucine, and aspartic acid (r = 0.47-0.56, P < 0.05) showed significant correlation to PhIP in chicken. Also a linear correlation was found to exist between PhIP (r = 0.87, P < 0.001) and MeIQx (r = 0.35, P < 0.01), and the molar ratio of creatin(in)e to glucose, respectively. Harman as co-mutagens was linearly correlated to the concentration of glucose (r = 0.65, P < 0.001). By contrast, norharman was not significant correlated to glucose levels.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Acetyl chloride solution, 1 M in methylene chloride
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Picric acid, moistened with water, ≥98%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Acetyl chloride, reagent grade, 98%
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Acetyl chloride, reagent grade, 98%
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
SAFC
L-Methionine
SAFC
L-Isoleucine
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)