Merck
  • Evaluation of the change in sphingolipids in the human multiple myeloma cell line U266 and gastric cancer cell line MGC-803 treated with arsenic trioxide.
Evaluation of the change in sphingolipids in the human multiple myeloma cell line U266 and gastric cancer cell line MGC-803 treated with arsenic trioxide.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-10-12)
Jianhua Zou, Xiaoqiong Ma, Guangji Zhang, Li Shen, Liting Zhou, Yu Yu, Fanfan Zhu, Zhe Chen
ABSTRACT
Arsenic trioxide (As2O3) has been found to display anticancer activity against many types of tumors and has been developed into an anticancer drug in clinical treatments. Sphingolipids are membrane lipids that participate in many signal transduction pathways. In this paper, the changes in sphingolipids of the human multiple myeloma cell line U266 and the gastric cancer cell line MGC-803 treated with arsenic trioxide were investigated using an HPLC-ESI-MS/MS method. Analytes were separated by an XBridge BEH C8 column used for Cer, HexCer, LacCer and SM chromatographic separation, and a Capcell PAK MG II C18 column was used for Sph, dhSph, S1P and dhS1P chromatographic separation and gradient elution with acetonitrile-water containing 0.1% formic acid as a mobile phase. A tandem mass spectrometer QTrap in SRM mode was employed in combination with RPLC as a detector for quantitative analysis. The ceramide/sphingolipid internal standard (IS) mixture was used to quantify the levels of sphingolipids. The distributions of sphingolipids were found to be different in the human multiple myeloma cell line U266 and the gastric cancer cell line MGC-803. Ceramide (Cer), hexosylceramide (HexCer) and dihexosylceramide (Hex2Cer) levels in U266 cell line are higher than those in MGC-803 cell line. Additionally, sphingomyelin (SM), sphingosine-1-phosphate (S1P) and sphinganine-1-phosphate (dhS1P) levels in the MGC-803 cell line are higher than those in the U266 cell line. When treated with arsenic trioxide (1-5μM iAs(III)(As(III) ions)), the levels of Hex2Cer in the human multiple myeloma cell line U266 decreased, and the levels of S1P and dhS1P in the human gastric cancer cell line MGC-803 decreased. The decrease of Hex2Cer, S1P and dhS1P in the human multiple myeloma cell line U266 and gastric cancer cell line MGC-803 were observed when the concentration of iAs(III) is 1.0μM. Therefore, arsenic trioxide exhibits anti-cancer activity by altering the sphingolipid pathway in the human multiple myeloma cell line U266 and the gastric cancer cell line MGC-803.
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, ≥98.0%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%