Merck
  • Reproductive performance in ewes fed varying levels of cut lucerne pasture around conception.
Reproductive performance in ewes fed varying levels of cut lucerne pasture around conception.
Animal reproduction science (2015-05-31)
S M Robertson, E H Clayton, B Morgan, M A Friend
ABSTRACT
Elevated intakes of protein and energy may increase embryo mortality, but it is not clear whether fresh lucerne (Medicago sativa) pasture poses a risk. A two-year pen study using oestrous synchronised and artificially inseminated Merino ewes (n=175 in 2013 and 215 in 2014) evaluated whether feeding freshly cut lucerne pasture (mean crude protein 19.7%, metabolisable energy 9.4MJ/kg DM) at maintenance or ad libitum during different periods around insemination altered reproductive performance in comparison with ewes fed a Control diet (mean crude protein 7.8%, metabolisable energy 9.0MJ/kg DM) of pelleted faba bean hulls and oat grain hulls at maintenance. The proportion of pregnant ewes carrying multiple fetuses was reduced (P=0.026) when ewes were fed lucerne ad libitum between days 0 and 17 after insemination compared with the Control diet (0.18 and 0.34, respectively), but not when ewes were fed lucerne ad libitum between days 0 and 7 after insemination (0.22). Reproductive performance, including the proportion of ewes pregnant and the proportion with multiple fetuses, was not different (P>0.05) when ewes were fed lucerne at maintenance between days 0 and 7 compared with the Control diet. While reproductive performance was similar when ewes were fed lucerne at maintenance between 0 and 17 days after artificial insemination compared with pellets at maintenance, fetal numbers per pregnant ewe were reduced by feeding lucerne ad libitum after insemination.
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture