Merck
  • Live births following Karyomapping of human blastocysts: experience from clinical application of the method.
Live births following Karyomapping of human blastocysts: experience from clinical application of the method.
Reproductive biomedicine online (2015-07-25)
Michalis Konstantinidis, Renata Prates, N-Neka Goodall, Jill Fischer, Victoria Tecson, Tsion Lemma, Bo Chu, Amy Jordan, Erin Armenti, Dagan Wells, Santiago Munné
ABSTRACT
The clinical application of a new, widely applicable method known as Karyomapping to carry out a total of 55 clinical cases of preimplantation genetic diagnosis (PGD) for single gene disorders is reported. Conventional polymerase chain reaction (PCR) testing was carried out in parallel to the new method for all cases. Clinical application of Karyomapping in this study resulted in three live births and nine clinical pregnancies out of 20 cases with a transfer. All in all, results presented in this study indicate that Karyomapping is a highly efficient, accurate and robust method for PGD of single gene disorders. Karyomapping can offer a more comprehensive assessment of the region of interest than conventional PCR analysis, allowing for more embryos to receive diagnosis (99.6% versus 96.8%), whereas its wide applicability reduces substantially the time that patients have to wait before starting their in vitro fertilization (IVF) cycle. Nonetheless, inclusion of elements of conventional PCR methodology, such as direct mutation detection, may be required in cases in which the gene of interest is in a region with reduced single nucleotide polymorphism (SNP) coverage (e.g. telomeric regions), when offering PGD for consanguineous couples, or in cases where no samples from additional family members are available.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)