Merck
  • Flavin-containing monooxygenase 3 as a potential player in diabetes-associated atherosclerosis.
Flavin-containing monooxygenase 3 as a potential player in diabetes-associated atherosclerosis.
Nature communications (2015-04-08)
Ji Miao, Alisha V Ling, Praveen V Manthena, Mary E Gearing, Mark J Graham, Rosanne M Crooke, Kevin J Croce, Ryan M Esquejo, Clary B Clish, David Vicent, Sudha B Biddinger
ABSTRACT
Despite the well-documented association between insulin resistance and cardiovascular disease, the key targets of insulin relevant to the development of cardiovascular disease are not known. Here, using non-biased profiling methods, we identify the enzyme flavin-containing monooxygenase 3 (Fmo3) to be a target of insulin. FMO3 produces trimethylamine N-oxide (TMAO), which has recently been suggested to promote atherosclerosis in mice and humans. We show that FMO3 is suppressed by insulin in vitro, increased in obese/insulin resistant male mice and increased in obese/insulin-resistant humans. Knockdown of FMO3 in insulin-resistant mice suppresses FoxO1, a central node for metabolic control, and entirely prevents the development of hyperglycaemia, hyperlipidemia and atherosclerosis. Taken together, these data indicate that FMO3 is required for FoxO1 expression and the development of metabolic dysfunction.
Sigma-Aldrich
MISSION® esiRNA, targeting human SREBF2
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Srebf2