Merck
  • Ru-assisted synthesis of Pd/Ru nanodendrites with high activity for ethanol electrooxidation.
Ru-assisted synthesis of Pd/Ru nanodendrites with high activity for ethanol electrooxidation.
Nanoscale (2015-07-03)
Ke Zhang, Duan Bin, Beibei Yang, Caiqin Wang, Fangfang Ren, Yukou Du
ABSTRACT
Due to the specific physical and chemical properties of a highly branched noble metal, the controllable synthesis has attracted much attention. This article reports the synthesis of Pd/Ru nanodendrites by a facile method using an oil bath in the presence of polyvinyl pyrrolidone, potassium bromide and ascorbic acid. The morphology, structure, and composition of the as-prepared catalysts were characterized by means of X-ray diffraction, X-ray photoelectron spectroscopy and transmission electron microscopy. In the electrochemical measurement, the as-prepared Pd7/Ru1 bimetallic nanodendrites provide a large electrochemically active surface area and exhibit high peak current density in the forward scan toward ethanol electrooxidation, which is nearly four times higher than those of a pure Pd catalyst. The as-prepared Pd7/Ru1 catalysts also exhibit significantly enhanced cycling stability toward ethanol oxidation in alkaline medium, which are mainly ascribed to the synergetic effect between Pd and Ru. This indicates that the Pd7/Ru1 catalysts should have great potential applications in direct ethanol fuel cells.
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Palladium(II) chloride solution, 5 wt. % in 10 wt. % HCl
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Palladium(II) chloride ChemBeads
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Palladium(II) chloride, ReagentPlus®, 99%
Sigma-Aldrich
2-Pyrrolidinone, ≥99%
Sigma-Aldrich
Palladium(II) chloride, ≥99.9%
Sigma-Aldrich
2-Pyrrolidinone, 99%
Sigma-Aldrich
Palladium(II) chloride, anhydrous, 59-60% palladium (Pd) basis
Sigma-Aldrich
Palladium(II) chloride, 99.995%
Sigma-Aldrich
Palladium(II) chloride, anhydrous, 60% Pd basis
Sigma-Aldrich
Potassium hydroxide, ≥99.95% trace metals basis