Merck
  • The use of a hydrogel matrix for controlled delivery of niacin to the gastrointestinal tract for treatment of hyperlipidemia.
The use of a hydrogel matrix for controlled delivery of niacin to the gastrointestinal tract for treatment of hyperlipidemia.
Physiological research (2015-10-09)
J Sirc, J Hrib, M Vetrik, R Hobzova, A Zak, B Stankova, O Slanar, R Hromadka, V Sandrikova, J Michalek
ABSTRACT
Hyperlipidemia treatment based on niacin requires gastrointestinal administration of relatively high doses. The recommended dietary allowance of niacin as vitamin B3 is 14 to 16 mg daily in adults, while the doses of niacin used in the treatment of hyperlipidemia are generally in the range of 1 to 3 g. Administration of such large doses requires a high concentration of the active compound in the tablet and proper control of the drug release. In this study, a hydrogel matrix based on poly(2-hydroxyethyl methacrylate) and polyvinylpyrrolidone was investigated as delivery vehicle for controlled NA release into the gastrointestinal environment. The prepared hydrogel matrices varied in used monomer and crosslinker types and concentrations. The content of NA in tablets was between 65-80 %. The release profiles of NA from tablets were examined under three different pH values (1, 4.5 and 6.8) over the time period of 30 h. The effects of the monomer ratio, the crosslinking of the polymer network, and the solubility of niacin during drug release under various pH are discussed. The results showed that the release time period can be achieved in a relatively wide range of time and can be adjusted according to the medical requirements.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
2-Hydroxyethyl methacrylate, contains ≤250 ppm monomethyl ether hydroquinone as inhibitor, 97%
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Supelco
Nicotinic acid, analytical standard
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Sigma-Aldrich
2-Hydroxyethyl methacrylate
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
Sigma-Aldrich
2-Hydroxyethyl methacrylate, ≥99%, contains ≤50 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Nicotinic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
1-Vinyl-2-pyrrolidinone, contains sodium hydroxide as inhibitor, ≥99%