Merck
  • Relationship of leptin with adiposity and inflammation and resistin with disease severity in psoriatic patients undergoing anti-TNF-alpha therapy.
Relationship of leptin with adiposity and inflammation and resistin with disease severity in psoriatic patients undergoing anti-TNF-alpha therapy.
Journal of the European Academy of Dermatology and Venereology : JEADV (2015-04-01)
T Pina, F Genre, R Lopez-Mejias, S Armesto, B Ubilla, V Mijares, T Dierssen-Sotos, M A Gonzalez-Lopez, M C Gonzalez-Vela, R Blanco, J L Hernández, J Llorca, M A Gonzalez-Gay
ABSTRACT
Altered secretion patterns of proinflammatory adipokines may influence the increased risk of cardiovascular mortality observed in patients with chronic inflammatory diseases. To determine whether two adipokines, leptin and resistin, correlate with metabolic syndrome features and disease severity in psoriatic patients who underwent anti-TNF-α therapy. Prospective study of consecutive non-diabetic patients with moderate-to-severe psoriasis who completed 6 months of therapy with anti-TNF-α- adalimumab. Patients with kidney disease, hypertension or body mass index ≥35 Kg/m(2) were excluded. Metabolic and clinical evaluation was performed at the onset of anti-TNF-α treatment and at month 6. Twenty-nine patients were assessed. A correlation between adiposity and leptin was observed (waist circumference and leptin levels after 6 months of therapy: r = 0.43; P = 0.030). Leptin concentration also correlated with blood pressure before adalimumab onset (systolic: r = 0.48; P = 0.013 and diastolic blood pressure: r = 0.50; P = 0.010 ). A marginally significant negative correlation between insulin sensitivity (QUICKI) and leptin levels was also observed. CRP levels correlated with leptin prior to the onset of adalimumab (r = 0.45; P = 0.020) and with resistin both before (r = 0.45; P = 0.020) and after 6 months of therapy (r = 0.55; P = 0.004). A positive association between parameters of disease activity such as BSA (r = 0.60; P = 0.001) and PASI (r = 0.63; P = 0.001) prior to the onset of adalimumab therapy and resistin concentrations was also disclosed. No significant changes in leptin and resistin concentrations following the 6-month treatment with adalimumab were seen. In patients with moderate-to-severe psoriasis leptin correlates with metabolic syndrome features and inflammation whereas resistin correlate with inflammation and disease severity.
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications