Merck
  • Comparison of the Odontogenic Differentiation Potential of Dental Follicle, Dental Papilla, and Cranial Neural Crest Cells.
Comparison of the Odontogenic Differentiation Potential of Dental Follicle, Dental Papilla, and Cranial Neural Crest Cells.
Journal of endodontics (2015-04-18)
Gang Chen, Qince Sun, Li Xie, Zongting Jiang, Lian Feng, Mei Yu, Weihua Guo, Weidong Tian
ABSTRACT
During tooth development, cells originating from the neural crest serve as precursors to the cells in the dental follicle and dental papilla. Therefore, the current study aimed to understand the associations of cranial neural crest cells (CNCCs), dental follicle cells (DFCs), and dental papilla cells (DPCs) by performing a parallel comparison to evaluate their odontogenic differentiation capacities. In this study, we harvested the 3 cells from C57/green fluorescent protein-positive mice or embryos and compared the cell morphology, surface antigens, microstructures, and gene and protein expression. Under the odontogenic microenvironments provided by treated dentin matrix, the odontogenic differentiations of the 3 cells were further compared in vitro and in vivo. The gene levels of DFCs in neurofilament, tubulin, and nestin were close to the DPCs, and in alkaline phosphatase, osteopontin, dentin matrix protein 1, and dentin sialophosphoprotein were the lowest in the 3 cells. However, Western blot results showed that DFCs possessed more similar protein profiles to CNCCs than DPCs, including collagen 1, transforming growth factor beta 1, osteopontin, neurofilament, and dentin matrix protein 1. Meanwhile, DFCs as 1 source of dental stem cells possessed high potency in odontogenic differentiation in vitro. Moreover, similar dentinlike tissues were observed in all 3 groups in vivo. CNCCs, DFCs, and DPCs possessed different biological characteristics in odontogenic differentiation.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)