Merck
  • Porphyrinic metal-organic framework as electrochemical probe for DNA sensing via triple-helix molecular switch.
Porphyrinic metal-organic framework as electrochemical probe for DNA sensing via triple-helix molecular switch.
Biosensors & bioelectronics (2015-05-08)
Pinghua Ling, Jianping Lei, Huangxian Ju
ABSTRACT
An electrochemical DNA sensor was developed based on the electrocatalysis of porphyrinic metal-organic framework (MOF) and triple-helix molecular switch for signal transduction. The streptavidin functionalized zirconium-porphyrin MOF (PCN-222@SA) was prepared as signal nanoprobe via covalent method and demonstrated high electrocatalysis for O2 reduction. Due to the large steric effect, the designed nanoprobe was blocked for the interaction with the biotin labeled triple-helix immobilized on the surface of glassy carbon electrode. In the presence of target DNA, the assistant DNA in triple-helix will hybridize with target DNA, resulting in the disassembly of triple-helix molecular. Consequently, the end biotin away from the electrode was ''activated'' for easy access to the signal nanoprobe, PCN-222@SA, on the basis of biotin-streptavidin biorecognition. The introduction of signal nanoprobe to a sensor surface led to a significantly amplified electrocatalytic current towards oxygen reduction. Integrating with DNA recycling amplification of Exonuclease III, the sensitivity of the biosensor was improved significantly with detection limit of 0.29 fM. Moreover, the present method has been successfully applied to detect DNA in complex serum matrix. This porphyrinic MOF-based strategy has promising application in the determination of various analytes for signal transduction and has great potential in bioassays.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Tetraethyl orthosilicate, ≥99.0% (GC)
Sigma-Aldrich
Tetraethyl orthosilicate, reagent grade, 98%
Sigma-Aldrich
Tetraethyl orthosilicate, 99.999% trace metals basis
Sigma-Aldrich
Tetraethyl orthosilicate, packaged for use in deposition systems
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Tetramethyl orthosilicate, ≥99%
Sigma-Aldrich
Tetramethyl orthosilicate, 98%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%