Merck
A battery-operated portable handheld electrospinning apparatus.
Nanoscale (2015-07-15)
Shi-Cong Xu, Chong-Chong Qin, Miao Yu, Rui-Hua Dong, Xu Yan, Hui Zhao, Wen-Peng Han, Hong-Di Zhang, Yun-Ze Long
ABSTRACT
Electrospinning (e-spinning) still has certain limitations in flexible practicability because its conventional setup is usually quite bulky and excessively dependent on a plug (electric supply). In this article, we report on a battery-operated e-spinning apparatus (BOEA) based on miniaturization and integration. The new device gets liberated from the conventional heavy power supply, achieves the tight integration of functional parts and can be operated by a single hand due to its small volume (10.5 × 5 × 3 cm(3)) and light weight (about 120 g). Different polymers such as polyvinylpyrrolidone (PVP), polycaprolactone (PCL), polystyrene (PS), poly(lactic acid) (PLA) and poly(vinylidene fluoride) (PVDF) were electrospun into fibers successfully, which confirms the stable performance and good real-time control capability of the apparatus. These results demonstrate that the BOEA could be potentially applied in many fields, especially in biomedical fields such as skin damage, wound healing, rapid hemostasis, etc.
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
2-Pyrrolidinone, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
2-Pyrrolidinone, 99%