Merck
  • Role of bone marrow mesenchymal stem cells in L-arg-induced acute pancreatitis: effects and possible mechanisms.
Role of bone marrow mesenchymal stem cells in L-arg-induced acute pancreatitis: effects and possible mechanisms.
International journal of clinical and experimental pathology (2015-07-21)
Guojian Yin, Guoyong Hu, Rong Wan, Ge Yu, Xiaofeng Cang, Jianbo Ni, Jie Xiong, Yanling Hu, Miao Xing, Yuting Fan, Wenqin Xiao, Lei Qiu, Shaofeng Wang, Xingpeng Wang
ABSTRACT
Mesenchymal stem cells (MSCs) have shown an obvious protective effect on systemic inflammation. The purpose of this study is to assess the effect and possible mechanism of bone marrow MSCs (bmMSCs) on acute pancreatitis (AP). BmMSCs of SD rats were isolated and cultured in vitro. L-Arginine-induced acute pancreatitis was used as AP model in vivo. Pancreatic injury was assessed by serum amylase, lipase, cytokines and pancreatic histology. RT-PCR was applied to investigate mRNA expression of pancreas tissue. Western-blot and immunohistochemistry (IHC) were applied to test the role of NF-κB p65 signaling pathway. Tracking and Positioning of CM-Dil labeled bmMSCs in vivo was further studied. Treatment with bmMSCs attenuated acute pancreatic injury and AP-associated lung injury obviously, with decreased serum IL-1β, IL-6, TNF-α, down-regulated expressions of IL-1α, IL-6, TNFα in pancreas tissue and reduced nuclear translocation of NF-κB p65 in AP. Localization of bmMSCs in vivo was due to being passively trapped in related organs, but not actively homing to inflammatory sites of pancreas during the early phase of AP. Taken together, the results showed that bmMSCs played a protective role in AP in many aspects, which might protect against experimental pancreatitis partly by regulating release of inflammatory cytokines by an exocrine secretion.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
Sigma-Aldrich
Selenous acid, 99.999% trace metals basis
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Selenous acid, 98%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
SAFC
L-Arginine
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O