Merck
  • Curcumin suppresses stem-like traits of lung cancer cells via inhibiting the JAK2/STAT3 signaling pathway.
Curcumin suppresses stem-like traits of lung cancer cells via inhibiting the JAK2/STAT3 signaling pathway.
Oncology reports (2015-09-24)
Lichuan Wu, Liangxia Guo, Yuehui Liang, Xu Liu, Lihe Jiang, Lisheng Wang
ABSTRACT
Tumor recurrence and drug resistance are the main obstacles blocking effective treatment of cancer patients. Cancer stem cells (CSCs) have been demonstrated to be highly related to tumor recurrence and drug resistance. Thus, eliminating CSCs may be an alternative for cancer therapy. Tumor sphere formation is a functional assay to enrich the CSC-like cells. In the present study, we tested the effects of curcumin on lung cancer stem-like cells and report that in addition to inhibition on the proliferation and colony formation of lung cancer cells, curcumin reduces tumor spheres of H460 cells. Moreover, by molecular docking analysis and tumor sphere assay we discover that curcumin was able to inhibit JAK2 activity and reduce tumor spheres via inhibiting the JAK2/STAT3 signaling pathway. In a lung cancer xenograft nude mouse model, curcumin strongly repressed tumor growth. These results imply curcumin may be a potential drug in lung CSC elimination and cancer therapy.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Stat3
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sigma-Aldrich
MISSION® esiRNA, targeting human STAT3