Merck
  • Intracellular delivery cellulose-based bionanogels with dual temperature/pH-response for cancer therapy.
Intracellular delivery cellulose-based bionanogels with dual temperature/pH-response for cancer therapy.
Colloids and surfaces. B, Biointerfaces (2015-06-30)
Yifen Wen, Jung Kwon Oh
ABSTRACT
Polysaccharide-based crosslinked nanogles (bionanogels) exhibiting multiple stimuli-responsive release of encapsulated therapeutics hold a great potential as tumor-targeting intracelluar durg delivery nanocarriers. Herein, we report the synthesis of monodisperse dual temperature/acidic pH-responsive bionanogels (DuR-BNGs) by aqueous crosslinking polymerization through temperature-induced self-association method. The DuR-BNGs have prolonged colloidal stability and negligible non-specific interactions with proteins. In response to acidic pH at higher temperature (above lower critical solution temperature), they exhibit synergistic release of anticancer drugs as a consequence of both acidic pH-sensitivity of carboxymethyl cellulose and temperature-induced volume change of grafted thermoresponsive copolymers. In vitro cell culture results suggest that new colloidally-stable DuR-BNG is a promising candidate promoting dual stimuli-responsive drug release for cancer therapy.
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ethylene oxide solution, 2.5-3.3 M in THF
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium hydrogen phthalate, BioXtra, ≥99.95%
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Potassium phthalate monobasic, ACS reagent, acidimetric standard
Sigma-Aldrich
5-(N,N-Dimethyl)amiloride hydrochloride
Sigma-Aldrich
Potassium phthalate monobasic, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Potassium phthalate monobasic, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
Ethylene oxide, ≥99.5%