Merck
  • Human Progenitor Cell Quantification After Xenotransplantation in Rat and Mouse Models by a Sensitive qPCR Assay.
Human Progenitor Cell Quantification After Xenotransplantation in Rat and Mouse Models by a Sensitive qPCR Assay.
Cell transplantation (2014-05-23)
Julie Prigent, Astrid Herrero, Jérôme Ambroise, Françoise Smets, Gisèle A Deblandre, Etienne M Sokal
ABSTRACT
Xenotransplantation of human cells in animal models is an essential tool for evaluation of safety and efficacy of cell-based products for therapeutic use. Sensitive and reproducible methods are needed to detect and quantify human cells engrafted into the host tissue either in the targeted organ or in undesired locations. We developed a robust quantitative polymerase chain reaction (qPCR) assay based on amplification of human AluYb8 repeats, to assess the number of human cells present in rat or mouse tissues after transplantation. Standard curves of mixed human/rodent DNA and mixed human/rodent cells have been performed to determine the limit of detection and linear range of the assay. Standard curves from DNA mixing differed significantly from standard curves from cell mixing. We show here that the AluYb8 qPCR assay is highly reproducible and is able to quantify human cells in a rodent cell matrix over a large linear range that extends from 50% to 0.01% human cells. Short-term in vivo studies showed that human cells could be quantified in mouse liver up to 7 days after intrasplenic transplantation and in rat liver 4 h after intrahepatic transplantation.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(±)-CPP, solid
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture