Merck
  • Versatile MoS2 Nanosheets in ITO-Free and Semi-transparent Polymer Power-generating Glass.
Versatile MoS2 Nanosheets in ITO-Free and Semi-transparent Polymer Power-generating Glass.
Scientific reports (2015-07-17)
Xiaotian Hu, Lie Chen, Licheng Tan, Yong Zhang, Lin Hu, Bing Xie, Yiwang Chen
ABSTRACT
Chemical exfoliated ultra-thin MoS2 nanosheets (NSs) with well 2D structure were demonstrated for interfacial layers and Ag nanowires composite transparent electrode in polymer solar cells (PSCs). The smooth and uniform n-type and p-type (after the plasma treatment) MoS2 NSs could improve fill factor of devices and light absorption in active layer. The optimized Ag nanowires-MoS2 NSs (AgNW-MoS2 NSs) transparent electrode presented a low sheet resistance of 9.8 Ω sq(-1), and the corresponding transmittance also exhibited a high value of 93.1% at 550 nm. As a result, ITO-free PSCs based on AgNW-MoS2 NSs/n-MoS2 NSs cathode and p-MoS2 NSs/Ag anode achieved a highest PCE of 8.72%. Furthermore, a high efficiency (6.55%), large area and low cost semi-transparent power-generating glass was obtained, after reducing the thickness of top Ag electrode from 100 nm to 30 nm. To our best knowledge, it is the highest performance for semi-transparent PSCs devices reported up to now. The novel semi-transparent power-generating glass showed good performance and color purity for commercial applications in the near future.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
n-Butyllithium solution, 2.5 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 2.0 M in cyclohexane
Sigma-Aldrich
n-Butyllithium solution, 1.6 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 10.7 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 2.7 M in heptane
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Hexane, anhydrous, 95%