Merck
  • Thalassemia Phenotypes and Genotypes in Taiwan: A Retrospective Study Based on Thalassemia Screening of Young Men for Military Conscription.
Thalassemia Phenotypes and Genotypes in Taiwan: A Retrospective Study Based on Thalassemia Screening of Young Men for Military Conscription.
Hemoglobin (2015-06-20)
Hsiao-Wei Lee, Shao-Min Han, Youngsen Yang, Tseng-Hsi Lin, Huey-En Tzeng, Kuang-Hsi Chang, Wen-Li Hwang, Chieh-Lin J Teng
ABSTRACT
Prenatal thalassemia studies from Taiwan show that one-third of fetuses with genetic abnormalities have β-thalassemia major (β-TM). However, the phenotypes and genotypes of adult thalassemia warrant further investigation. From September 2006 to April 2014, 741 male candidates drafted for military service with mean corpuscular volume (MCV) <80 fL and serum ferritin >20 µg/L were analyzed. The results showed that the detection rates of α- and β-thalassemia (α- an β-thal) were 50.20% (372/741) and 49.12% (364/741), respectively. Only five patients (0.67%) were diagnosed with both α- and β-thal. The - -(SEA)/αα mutation was found in 76.88% (286/372) of α-thal patients. Heterozygous mutations in IVS-II-654 (C > T) and codons 41/42 (-TCTT) accounted for 55.77% (203/364) of β-thal cases. The leukocyte counts for α- and β-thal were 6241.74 ± 1552.99 and 6622.87 ± 1814.41 × 10(9)/L, respectively (p = 0.007). The α-thal patients had lower red blood cell (RBC) mass (5.85 ± 0.44 × 10(12)/L vs. 6.09 ± 0.45 × 10(12)/L; p < 0.001) and higher hemoglobin (Hb) (12.82 ± 0.72 vs. 12.35 ± 0.71 g/dL; p < 0.001) than β-thal patients. Mean serum ferritin values were 169.67 and 241.36 µg/L, respectively, in α- and β-thal patients (p < 0.001), indicating more profound ineffective erythropoiesis in β-thal. Only four of the 741 patients underwent further hematological follow-up. Our study suggests that iron overload might be a potential problem in β-thal patients; therefore, regular follow-up is highly recommended.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M