Merck
  • MicroRNA-144 suppresses osteosarcoma growth and metastasis by targeting ROCK1 and ROCK2.
MicroRNA-144 suppresses osteosarcoma growth and metastasis by targeting ROCK1 and ROCK2.
Oncotarget (2015-04-29)
Wei Wang, Xin Zhou, Min Wei
ABSTRACT
Osteosarcoma (OS) is the most common primary tumor of bone. MicroRNAs (miRNAs) are a class of endogenously expressed small non-coding RNAs that are strongly implicated in cancerous processes. However, our current understanding of the biological role of miRNAs in OS remains incomplete. In the present study, miR-144 was markedly downregulated in OS cell lines and clinical specimens. Low-level expression of miR-144 was significantly associated with distant metastasis and poor prognosis. Functional studies demonstrated that ectopic expression of miR-144 suppresses tumor cell proliferation and metastasis in vitro as well as in vivo. Furthermore, we identified Rho-associated kinases 1 and 2 (ROCK1 and ROCK2) as direct targets for miR-144 binding, resulting in suppression of their expression. Exogenous expression of ROCK1 or ROCK2 in 143B-miR-144 cells partially restored miR-144-inhibited cell proliferation and invasion. In clinical OS specimens, ROCK1 and ROCK2 levels were elevated, relative to that in paired normal bone tissues, and inversely correlated with miR-144 expression. Taken together, miR-144 suppresses OS progression by directly downregulating ROCK1 and ROCK2 expression, and may be a promising therapeutic target for OS.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rock1
Sigma-Aldrich
MISSION® esiRNA, targeting human ROCK2
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
MISSION® esiRNA, targeting human ROCK1 (2)