Merck
  • Identification of Ellagitannins and Flavonoids from Eugenia brasilienses Lam. (Grumixama) by HPLC-ESI-MS/MS.
Identification of Ellagitannins and Flavonoids from Eugenia brasilienses Lam. (Grumixama) by HPLC-ESI-MS/MS.
Journal of agricultural and food chemistry (2015-05-21)
Luciane de Lira Teixeira, Fabiano Cleber Bertoldi, Franco Maria Lajolo, Neuza Mariko Aymoto Hassimotto
ABSTRACT
The grumixama (Eugenia brasiliensis Lam.), also known as Brazilian cherry, is a fruit native to Brazil. This study identified the flavonoids in the flesh and seeds and ellagitannin in the flesh of purple and yellow varieties. The physicochemical characteristics and antioxidant capacity of these fruits were also evaluated. Anthocyanins and flavonols were found in high levels in the flesh of purple (32-180 mg 100 g(-1) FW) and yellow grumixama (13-41 mg 100 g(-1) FW), respectively. The major flavonoids identified were cyanidin 3-glucoside and quercetin aglycone. Furthermore, ellagitannins were found in high levels in the flesh of purple (82-243 mg ellagic acid equiv 100 g(-1) FW) and yellow grumixama (92 mg ellagic acid equiv 100 g(-1) FW) and seeds (2220-2905 mg ellagic acid equiv 100 g(-1) FW). The ellagitannin profiles of both varieties were first characterized in which pedunculagin isomers, strictinin isomers, and ellagic acid galloyl hexoside were the major ellagitannins identified. In summary, both varieties of the grumixama fruit as well as the seeds could be good sources of bioactive compounds, mainly ellagitannins.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Fluorescein, suitable for fluorescence, free acid
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
2,2′-Azobis(2-methylpropionamidine) dihydrochloride, powder or granules, 97%
Sigma-Aldrich
CPNQ, ≥98% (HPLC), solid
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%