Merck
  • Conformational transitions in the glycine-bound GluN1 NMDA receptor LBD via single-molecule FRET.
Conformational transitions in the glycine-bound GluN1 NMDA receptor LBD via single-molecule FRET.
Biophysical journal (2015-07-15)
David R Cooper, Drew M Dolino, Henriette Jaurich, Bo Shuang, Swarna Ramaswamy, Caitlin E Nurik, Jixin Chen, Vasanthi Jayaraman, Christy F Landes
ABSTRACT
The N-methyl-D-aspartate receptor (NMDAR) is a member of the glutamate receptor family of proteins and is responsible for excitatory transmission. Activation of the receptor is thought to be controlled by conformational changes in the ligand binding domain (LBD); however, glutamate receptor LBDs can occupy multiple conformations even in the activated form. This work probes equilibrium transitions among NMDAR LBD conformations by monitoring the distance across the glycine-bound LBD cleft using single-molecule Förster resonance energy transfer (smFRET). Recent improvements in photoprotection solutions allowed us to monitor transitions among the multiple conformations. Also, we applied a recently developed model-free algorithm called "step transition and state identification" to identify the number of states, their smFRET efficiencies, and their interstate kinetics. Reversible interstate conversions, corresponding to transitions among a wide range of cleft widths, were identified in the glycine-bound LBD, on much longer timescales compared to channel opening. These transitions were confirmed to be equilibrium in nature by shifting the distribution reversibly via denaturant. We found that the NMDAR LBD proceeds primarily from one adjacent smFRET state to the next under equilibrium conditions, consistent with a cleft-opening/closing mechanism. Overall, by analyzing the state-to-state transition dynamics and distributions, we achieve insight into specifics of long-lived LBD equilibrium structural dynamics, as well as obtain a more general description of equilibrium folding/unfolding in a conformationally dynamic protein. The relationship between such long-lived LBD dynamics and channel function in the full receptor remains an open and interesting question.
Sigma-Aldrich
Guanidine hydrochloride, ≥99% (titration), organic base and chaeotropic agent
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
Sigma-Aldrich
Guanidine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99%
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Guanidine hydrochloride, Molecular Biology, ≥99%
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Maleimide, 99%
SAFC
Isopropyl β-D-1-thiogalactopyranoside
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
SAFC
Glycine
SAFC
L-Threonine
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%